Product Details

SNP ID
rs112543324
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41879017 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGGTGCCAGGCCCTGTGGATGC[A/G]CAAGGTCCCAGCATCATTGATGGTG
Phenotype
MIM: 601855 MIM: 112205 MIM: 603474
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF1 PubMed Links

Gene Details

Gene
ARHGEF1
Gene Name
Rho guanine nucleotide exchange factor 1
There are no transcripts associated with this gene.

Gene
CD79A
Gene Name
CD79a molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001783.3 292 Missense Mutation CAC,CGC H36R NP_001774.1
NM_021601.3 292 Missense Mutation CAC,CGC H36R NP_067612.1
Gene
MIR6797
Gene Name
microRNA 6797
There are no transcripts associated with this gene.

Gene
RPS19
Gene Name
ribosomal protein S19
There are no transcripts associated with this gene.

View Full Product Details