Product Details

SNP ID
rs111707960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42232984 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACTTTGAAAGCGGATGGATGTTA[C/T]TGATCCCTGGATAACTCTTCAAATC
Phenotype
MIM: 606784 MIM: 614387
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GSK3A PubMed Links

Gene Details

Gene
GSK3A
Gene Name
glycogen synthase kinase 3 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019884.2 Intron NP_063937.2
Gene
ZNF526
Gene Name
zinc finger protein 526
There are no transcripts associated with this gene.

View Full Product Details