Product Details

SNP ID
rs112235686
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43071877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTGTGCCGGTGGGTTAGAGTTCG[C/T]GAAGCAAGACAAGTAGAGGTTATCT
Phenotype
MIM: 176391
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSG2 PubMed Links

Gene Details

Gene
PSG2
Gene Name
pregnancy specific beta-1-glycoprotein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031246.3 Intron NP_112536.2

View Full Product Details