Product Details

SNP ID
rs113072411
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43586845 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGTGAGGTAGATGCCATTTGTA[A/G]AGTGACTTGCCCAAGTCAGTGGAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IRGQ PubMed Links

Gene Details

Gene
IRGQ
Gene Name
immunity related GTPase Q
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007561.2 7288 UTR 3 NP_001007562.1
XM_005258515.3 7288 UTR 3 XP_005258572.1
Gene
PINLYP
Gene Name
phospholipase A2 inhibitor and LY6/PLAUR domain containing
There are no transcripts associated with this gene.

Gene
ZNF576
Gene Name
zinc finger protein 576
There are no transcripts associated with this gene.

View Full Product Details