Product Details

SNP ID
rs113478925
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46040391 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTCCACTCCCCGCACCTGGGCGC[C/T]GGCTGGCATAGCCACAGTCTAAGAT
Phenotype
MIM: 610547
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGFL4 PubMed Links

Gene Details

Gene
IGFL4
Gene Name
IGF like family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002923.1 150 Silent Mutation CCA,CCG P32P NP_001002923.1

View Full Product Details