Product Details

SNP ID
rs114886257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46410699 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTAAGTGTGCAGGCAAAAAAGAGA[A/T]GGAAAAAGGAGTCAGTTTCTCCCCT
Phenotype
MIM: 614145
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC8 PubMed Links

Gene Details

Gene
CCDC8
Gene Name
coiled-coil domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032040.4 2964 UTR 3 NP_114429.2

View Full Product Details