Product Details
- SNP ID
-
rs115702391
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:47781109 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCTCCCCTACCCCCTTTCCTCAGT[A/G]CGGCGAGGGAACTCCCCAGGCCACC
- Phenotype
-
MIM: 603235
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SEPW1
PubMed Links
Gene Details
- Gene
- SEPW1
- Gene Name
- selenoprotein W, 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003009.2 |
311 |
Missense Mutation |
|
|
NP_003000.1 |
View Full Product Details