Product Details

SNP ID
rs117085958
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48552811 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGTGTTCTAGGGCACTTGGGGC[A/G]GAGAGGGCAGGCTCTGGACTCTTCA
Phenotype
MIM: 604125
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SULT2B1 PubMed Links

Gene Details

Gene
SULT2B1
Gene Name
sulfotransferase family 2B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004605.2 Intron NP_004596.2
NM_177973.1 Intron NP_814444.1

View Full Product Details