Product Details

SNP ID
rs115519627
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49069448 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCATACAAACTTAACCCAATGTGA[C/T]CTAGAAAGCTCAAATTCTACCACTG
Phenotype
MIM: 176268 MIM: 162662
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNA7 PubMed Links

Gene Details

Gene
KCNA7
Gene Name
potassium voltage-gated channel subfamily A member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031886.2 2342 UTR 3 NP_114092.2
Gene
NTF4
Gene Name
neurotrophin 4
There are no transcripts associated with this gene.

View Full Product Details