Product Details

SNP ID
rs117745006
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49087657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGTCATCTCTTAGGTGCATCAGA[A/G]CACACCTGAGAAAAGCTGGCACACA
Phenotype
MIM: 180740
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNRNP70 PubMed Links

Gene Details

Gene
SNRNP70
Gene Name
small nuclear ribonucleoprotein U1 subunit 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301069.1 Intron NP_001287998.1
NM_003089.5 Intron NP_003080.2
XM_005259178.1 Intron XP_005259235.1
XM_011527240.2 Intron XP_011525542.2
XM_011527241.2 Intron XP_011525543.1

View Full Product Details