Product Details

SNP ID
rs112619445
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49117853 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGCTGTCTGTGTTGGGCCCTGC[A/T]GAGCGTTGAGGGTGAGCAGCATGAG
Phenotype
MIM: 612331 MIM: 603144 MIM: 180740
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C19orf73 PubMed Links

Gene Details

Gene
C19orf73
Gene Name
chromosome 19 open reading frame 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018111.2 Intron NP_060581.2
Gene
LIN7B
Gene Name
lin-7 homolog B, crumbs cell polarity complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308419.1 Intron NP_001295348.1
NM_022165.2 Intron NP_071448.1
XM_006723323.3 Intron XP_006723386.1
XM_017027131.1 Intron XP_016882620.1
Gene
PPFIA3
Gene Name
PTPRF interacting protein alpha 3
There are no transcripts associated with this gene.

Gene
SNRNP70
Gene Name
small nuclear ribonucleoprotein U1 subunit 70
There are no transcripts associated with this gene.

View Full Product Details