Product Details

SNP ID
rs112714381
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49807463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCTGATCCCAAGTTCTGAACGC[A/G]TGGGGTCTCTGAACACAGGTTTTGG
Phenotype
MIM: 601026 MIM: 610622
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AP2A1 PubMed Links

Gene Details

Gene
AP2A1
Gene Name
adaptor related protein complex 2 alpha 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014203.2 Intron NP_055018.2
NM_130787.2 Intron NP_570603.2
XM_011526556.2 Intron XP_011524858.1
XM_011526557.2 Intron XP_011524859.1
Gene
FUZ
Gene Name
fuzzy planar cell polarity protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171937.1 Intron NP_001165408.1
NM_025129.4 Intron NP_079405.2
XM_006723399.3 Intron XP_006723462.1
XM_011527339.1 Intron XP_011525641.1
XM_011527340.1 Intron XP_011525642.1
XM_011527341.2 Intron XP_011525643.1
XM_011527342.1 Intron XP_011525644.1
XM_011527343.1 Intron XP_011525645.1
XM_011527345.1 Intron XP_011525647.1
XM_011527346.1 Intron XP_011525648.1
XM_011527347.1 Intron XP_011525649.1
XM_017027319.1 Intron XP_016882808.1
XM_017027320.1 Intron XP_016882809.1
XM_017027321.1 Intron XP_016882810.1
XM_017027322.1 Intron XP_016882811.1
XM_017027323.1 Intron XP_016882812.1

View Full Product Details