Product Details

SNP ID
rs112796514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49951945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCCTGCTCAGCTGCTGCCCTCTT[A/G]CGAGCTTCCTTCCTGCAGATCTTCA
Phenotype
MIM: 607157
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SIGLEC11 PubMed Links

Gene Details

Gene
SIGLEC11
Gene Name
sialic acid binding Ig like lectin 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135163.1 1760 Intron NP_001128635.1
NM_052884.2 1760 Intron NP_443116.2
XM_005258476.3 1760 Silent Mutation CGC,CGT R576R XP_005258533.2
XM_005258477.3 1760 Intron XP_005258534.2

View Full Product Details