Product Details

SNP ID
rs112856426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50771366 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGTCCATGCCAACCGGCCCAAG[A/G]GGCTGCTGCTGGTGCTGGTGAGCGC
Phenotype
MIM: 603195
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR32 PubMed Links

Gene Details

Gene
GPR32
Gene Name
G protein-coupled receptor 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001506.2 903 Missense Mutation AGG,GGG R256G NP_001497.1
Gene
LOC105372440
Gene Name
uncharacterized LOC105372440
There are no transcripts associated with this gene.

View Full Product Details