Product Details

SNP ID
rs111689449
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50854966 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGCTGTGTCACCATGTGGGTCC[C/T]GGTTGTCTTCCTCACCCTGTCCGTG
Phenotype
MIM: 176820
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLK3 PubMed Links

Gene Details

Gene
KLK3
Gene Name
kallikrein related peptidase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001030047.1 52 Missense Mutation CCG,CTG P4L NP_001025218.1
NM_001030048.1 52 Missense Mutation CCG,CTG P4L NP_001025219.1
NM_001648.2 52 Missense Mutation CCG,CTG P4L NP_001639.1
Gene
LOC105372441
Gene Name
uncharacterized LOC105372441
There are no transcripts associated with this gene.

View Full Product Details