Product Details
- SNP ID
-
rs112865236
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:51022632 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGCTTTCGGGTGATCGCACACGG[A/G]TCCTGGCCCCAGGAGATAATGCCTT
- Phenotype
-
MIM: 602673
MIM: 604434
MIM: 605539
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KLK10
PubMed Links
Gene Details
- Gene
- KLK10
- Gene Name
- kallikrein related peptidase 10
There are no transcripts associated with this gene.
- Gene
- KLK11
- Gene Name
- kallikrein related peptidase 11
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001136032.2 |
795 |
Silent Mutation |
GAC,GAT |
D222D |
NP_001129504.1 |
NM_001167605.1 |
795 |
Silent Mutation |
GAC,GAT |
D247D |
NP_001161077.1 |
NM_006853.2 |
795 |
Silent Mutation |
GAC,GAT |
D222D |
NP_006844.1 |
NM_144947.1 |
795 |
Silent Mutation |
GAC,GAT |
D254D |
NP_659196.1 |
XM_005258439.3 |
795 |
Silent Mutation |
GAC,GAT |
D113D |
XP_005258496.1 |
XM_011526369.1 |
795 |
Silent Mutation |
GAC,GAT |
D279D |
XP_011524671.1 |
XM_011526370.2 |
795 |
Silent Mutation |
GAC,GAT |
D247D |
XP_011524672.1 |
XM_011526371.2 |
795 |
Silent Mutation |
GAC,GAT |
D247D |
XP_011524673.1 |
XM_011526372.2 |
795 |
Silent Mutation |
GAC,GAT |
D247D |
XP_011524674.1 |
XM_011526373.1 |
795 |
Silent Mutation |
GAC,GAT |
D247D |
XP_011524675.1 |
- Gene
- KLK12
- Gene Name
- kallikrein related peptidase 12
There are no transcripts associated with this gene.
View Full Product Details