Product Details

SNP ID
rs113751217
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51098874 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGGTCCAGCACCGCGGACGGCCG[C/G]GCCGCCTCCAGGCGCTTGAGCAGGT
Phenotype
MIM: 612694
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CTU1 PubMed Links

Gene Details

Gene
CTU1
Gene Name
cytosolic thiouridylase subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145232.3 839 Silent Mutation GCC,GCG A258A NP_660275.2

View Full Product Details