Product Details

SNP ID
rs112875957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53138957 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCATGGTATTCTGGTTTTTCTGA[C/T]CAAGCTCTCCACTCTAATAACTGTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF347 PubMed Links

Gene Details

Gene
ZNF347
Gene Name
zinc finger protein 347
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172674.1 4301 UTR 3 NP_001166145.1
NM_001172675.1 4301 UTR 3 NP_001166146.1
NM_032584.2 4301 UTR 3 NP_115973.2
XM_005259335.4 4301 UTR 3 XP_005259392.1
XM_005259336.4 4301 UTR 3 XP_005259393.1
XM_017027384.1 4301 UTR 3 XP_016882873.1
XM_017027385.1 4301 UTR 3 XP_016882874.1
XM_017027386.1 4301 UTR 3 XP_016882875.1
Gene
ZNF415
Gene Name
zinc finger protein 415
There are no transcripts associated with this gene.

View Full Product Details