Product Details

SNP ID
rs115941539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55364881 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGGCGGCTGGGTGGCGTTCTATC[C/T]ACAGATGCACCATGTTGCTTAACCC
Phenotype
MIM: 147681
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM71E2 PubMed Links

Gene Details

Gene
FAM71E2
Gene Name
family with sequence similarity 71 member E2
There are no transcripts associated with this gene.

Gene
IL11
Gene Name
interleukin 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000641.3 1879 UTR 3 NP_000632.1
NM_001267718.1 1879 UTR 3 NP_001254647.1

View Full Product Details