Product Details

SNP ID
rs113580964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56440341 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGAAGATCATATATCCTCATAT[C/T]CGTGGAGATGGTAAAGTAAGGTTTC
Phenotype
MIM: 611024
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF667 PubMed Links

Gene Details

Gene
ZNF667
Gene Name
zinc finger protein 667
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321355.1 2843 UTR 3 NP_001308284.1
NM_001321356.1 2843 UTR 3 NP_001308285.1
NM_022103.3 2843 UTR 3 NP_071386.3
XM_011527208.1 2843 UTR 3 XP_011525510.1
XM_011527209.1 2843 UTR 3 XP_011525511.1

View Full Product Details