Product Details
- SNP ID
-
rs117078524
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:56570276 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTTTCTCACTACTTCTTTTTCTCC[C/T]CAGCTCTACAATCCCAGAGTAAAGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF470
PubMed Links
Gene Details
- Gene
- ZNF470
- Gene Name
- zinc finger protein 470
View Full Product Details