Product Details

SNP ID
rs112107510
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57506414 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGAGCAGAGTGCTTCTGTAGAA[A/G]TGCCCAGTTCAAACGTTCAGCAACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF773 PubMed Links

Gene Details

Gene
ZNF773
Gene Name
zinc finger protein 773
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304334.1 543 Missense Mutation ATG,GTG M106V NP_001291263.1
NM_001304335.1 543 Intron NP_001291264.1
NM_001304336.1 543 UTR 3 NP_001291265.1
NM_001304337.1 543 Intron NP_001291266.1
NM_198542.2 543 Missense Mutation ATG,GTG M107V NP_940944.1

View Full Product Details