Product Details

SNP ID
rs111675758
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57605746 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCAGAGAGCGTTTCTGTGGAAG[A/C]ACTGTCACAGGGCAGGACTCCAAAG
Phenotype
MIM: 604076
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF134 PubMed Links

Gene Details

Gene
ZNF134
Gene Name
zinc finger protein 134
There are no transcripts associated with this gene.

Gene
ZNF530
Gene Name
zinc finger protein 530
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321981.1 441 Missense Mutation GAA,GCA E41A NP_001308910.1
NM_020880.4 441 Missense Mutation GAA,GCA E74A NP_065931.3
XM_006723194.1 441 Missense Mutation GAA,GCA E74A XP_006723257.1
XM_011526919.2 441 Missense Mutation GAA,GCA E73A XP_011525221.1
XM_011526921.2 441 Missense Mutation GAA,GCA E41A XP_011525223.1

View Full Product Details