Product Details
- SNP ID
-
rs113591395
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:58063456 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTCTCCCTGAGCAGGACATTGGTT[A/G]CCGAAGCCGAATGTCATCTCCCTGC
- Phenotype
-
MIM: 604077
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF135
PubMed Links
Gene Details
- Gene
- ZNF135
- Gene Name
- zinc finger protein 135
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001164527.1 |
95 |
Intron |
|
|
NP_001157999.1 |
NM_001164529.1 |
95 |
Silent Mutation |
TTA,TTG |
L69L |
NP_001158001.1 |
NM_001164530.1 |
95 |
Silent Mutation |
TTA,TTG |
L69L |
NP_001158002.1 |
NM_001289401.1 |
95 |
Silent Mutation |
TTA,TTG |
L57L |
NP_001276330.1 |
NM_001289402.1 |
95 |
Silent Mutation |
TTA,TTG |
L15L |
NP_001276331.1 |
NM_003436.3 |
95 |
Silent Mutation |
TTA,TTG |
L57L |
NP_003427.3 |
NM_007134.1 |
95 |
Silent Mutation |
TTA,TTG |
L69L |
NP_009065.1 |
XM_005259211.1 |
95 |
Silent Mutation |
TTA,TTG |
L69L |
XP_005259268.1 |
XM_006723362.3 |
95 |
Silent Mutation |
TTA,TTG |
L57L |
XP_006723425.1 |
XM_006723363.3 |
95 |
Silent Mutation |
TTA,TTG |
L57L |
XP_006723426.1 |
XM_017027240.1 |
95 |
Silent Mutation |
TTA,TTG |
L57L |
XP_016882729.1 |
XM_017027241.1 |
95 |
Silent Mutation |
TTA,TTG |
L15L |
XP_016882730.1 |
XM_017027242.1 |
95 |
Missense Mutation |
TAC,TGC |
Y27C |
XP_016882731.1 |
- Gene
- ZSCAN1
- Gene Name
- zinc finger and SCAN domain containing 1
There are no transcripts associated with this gene.
View Full Product Details