Product Details

SNP ID
rs114025965
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:32677721 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGCACACTTGCCTGCGGAAAAGG[A/G]CTCTCCCCAGCCACCCGGAGATGGG
Phenotype
MIM: 607814
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD27 PubMed Links

Gene Details

Gene
ANKRD27
Gene Name
ankyrin repeat domain 27
There are no transcripts associated with this gene.

Gene
RGS9BP
Gene Name
regulator of G-protein signaling 9 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207391.2 2315 UTR 3 NP_997274.2

View Full Product Details