Product Details

SNP ID
rs113061036
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:34759141 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTTCTCTCCAGTGTGAGTTCTCA[C/T]GTGCTGAGTTAAAGCAAAGTTGTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF599 PubMed Links

Gene Details

Gene
ZNF599
Gene Name
zinc finger protein 599
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007248.2 2048 Missense Mutation ATG,GTG M554V NP_001007249.1
XM_017026369.1 2048 Missense Mutation ATG,GTG M517V XP_016881858.1

View Full Product Details