Product Details

SNP ID
rs112627820
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35033740 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCACATTGAGGTAGTGGACAAAG[C/G]TGAGTCGGGTGCTGCCTGCCCCTTT
Phenotype
MIM: 142440 MIM: 600235
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
GRAMD1A PubMed Links

Gene Details

Gene
GRAMD1A
Gene Name
GRAM domain containing 1A
There are no transcripts associated with this gene.

Gene
HPN
Gene Name
hepsin
There are no transcripts associated with this gene.

Gene
SCN1B
Gene Name
sodium voltage-gated channel beta subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037.4 582 Intron NP_001028.1
NM_001321605.1 582 Intron NP_001308534.1
NM_199037.4 582 Missense Mutation GCT,GGT A150G NP_950238.1

View Full Product Details