Product Details

SNP ID
rs111275297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35668638 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGGTGCCGCACTCTGCCGCCGCC[A/G]GTCCATGGTCCTCACGGTGAGACTC
Phenotype
MIM: 124089 MIM: 611557
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COX6B1 PubMed Links

Gene Details

Gene
COX6B1
Gene Name
cytochrome c oxidase subunit 6B1
There are no transcripts associated with this gene.

Gene
UPK1A
Gene Name
uroplakin 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281443.1 296 Missense Mutation CAG,CGG Q90R NP_001268372.1
NM_007000.3 296 Missense Mutation CAG,CGG Q90R NP_008931.1
XM_017026225.1 296 Missense Mutation CAG,CGG Q91R XP_016881714.1
Gene
UPK1A-AS1
Gene Name
UPK1A antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details