Product Details

SNP ID
rs113554294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36039534 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTTGTGACCGCTCAGGAGTTGGC[A/G]CAGGGGCCAGGGGGGTCAGGAGCAT
Phenotype
MIM: 607382 MIM: 612536
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLIP3 PubMed Links

Gene Details

Gene
CLIP3
Gene Name
CAP-Gly domain containing linker protein 3
There are no transcripts associated with this gene.

Gene
LOC101927572
Gene Name
uncharacterized LOC101927572
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290056.1 815 Intron NP_001276985.1
Gene
THAP8
Gene Name
THAP domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152658.2 815 Missense Mutation GCG,GTG A154V NP_689871.1
XM_005258623.2 815 Missense Mutation GCG,GTG A111V XP_005258680.1
XM_011526604.2 815 Missense Mutation GCG,GTG A111V XP_011524906.1

View Full Product Details