Product Details

SNP ID
rs113215509
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36393158 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTTTCCAGCATTCCTTACATTC[A/G]TAAGGTTTTTCACCAGTATGAATTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZFP82 PubMed Links

Gene Details

Gene
ZFP82
Gene Name
ZFP82 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321917.1 1443 Silent Mutation TAA,TAG *395* NP_001308846.1
NM_001321918.1 1443 Intron NP_001308847.1
NM_001321919.1 1443 Intron NP_001308848.1
NM_133466.3 1443 Silent Mutation TAA,TAG *394* NP_597723.1

View Full Product Details