Product Details

SNP ID
rs115891922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36607571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCAGGACTGTTTCTTTTTCCAAA[A/C]GAGGAGCTCAAAAGAGAAAGTTCAT
Phenotype
MIM: 609516
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF382 PubMed Links

Gene Details

Gene
ZNF382
Gene Name
zinc finger protein 382
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256838.1 63 UTR 5 NP_001243767.1
NM_032825.4 63 UTR 5 NP_116214.2
Gene
ZNF529
Gene Name
zinc finger protein 529
There are no transcripts associated with this gene.

View Full Product Details