Product Details

SNP ID
rs117775896
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1614336 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCTCCCGGGTCTGGTTTCTTCCC[A/G]CAAGCCCTGACGGGGGGCTTTGGGG
Phenotype
MIM: 147141 MIM: 609711
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TCF3 PubMed Links

Gene Details

Gene
TCF3
Gene Name
transcription factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136139.2 Intron NP_001129611.1
NM_003200.3 Intron NP_003191.1
XM_006722852.3 Intron XP_006722915.1
XM_006722853.3 Intron XP_006722916.1
XM_006722855.3 Intron XP_006722918.1
XM_006722856.3 Intron XP_006722919.1
XM_006722857.3 Intron XP_006722920.1
XM_006722858.3 Intron XP_006722921.1
XM_011528215.2 Intron XP_011526517.1
XM_011528216.2 Intron XP_011526518.1
XM_011528217.2 Intron XP_011526519.1
XM_011528219.2 Intron XP_011526521.1
XM_011528220.2 Intron XP_011526522.1
XM_011528221.2 Intron XP_011526523.1
XM_011528223.2 Intron XP_011526525.1
XM_011528225.2 Intron XP_011526527.1
XM_011528226.2 Intron XP_011526528.1
XM_011528227.2 Intron XP_011526529.1
XM_017027177.1 Intron XP_016882666.1
XM_017027178.1 Intron XP_016882667.1
XM_017027179.1 Intron XP_016882668.1
XM_017027180.1 Intron XP_016882669.1
XM_017027181.1 Intron XP_016882670.1
XM_017027182.1 Intron XP_016882671.1
XM_017027183.1 Intron XP_016882672.1
XM_017027184.1 Intron XP_016882673.1
Gene
UQCR11
Gene Name
ubiquinol-cytochrome c reductase, complex III subunit XI
There are no transcripts associated with this gene.

View Full Product Details