Product Details

SNP ID
rs114346119
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2851487 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCAGATGATGAAACTCAATTTAA[G/T]GCCAGTGGGTCAGTTTCTCAGCAGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF555 PubMed Links

Gene Details

Gene
ZNF555
Gene Name
zinc finger protein 555
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172775.1 291 Silent Mutation AAG,AAT K50N NP_001166246.1
NM_152791.4 291 Silent Mutation AAG,AAT K50N NP_690004.4
XM_011527716.2 291 Silent Mutation AAG,AAT K52N XP_011526018.1
XM_017026375.1 291 Silent Mutation AAG,AAT K52N XP_016881864.1

View Full Product Details