Product Details

SNP ID
rs112180438
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4231074 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACCTATTTATTCCGAAAGCCTTT[A/T]TTAGGCACCTACCATGTACCTGGTG
Phenotype
MIM: 605816
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD24 PubMed Links

Gene Details

Gene
ANKRD24
Gene Name
ankyrin repeat domain 24
There are no transcripts associated with this gene.

Gene
EBI3
Gene Name
Epstein-Barr virus induced 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005755.2 Intron NP_005746.2
XM_011527619.2 Intron XP_011525921.1

View Full Product Details