Product Details

SNP ID
rs115919311
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5589209 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTGCTCACCATGGGTGAGTTTCC[C/T]GTTTACACTCCCGAGGTATTTACCA
Phenotype
MIM: 608066
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SAFB2 PubMed Links

Gene Details

Gene
SAFB2
Gene Name
scaffold attachment factor B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014649.2 Intron NP_055464.1
XM_011528449.2 Intron XP_011526751.1
XM_011528450.2 Intron XP_011526752.1
XM_017027520.1 Intron XP_016883009.1
XM_017027521.1 Intron XP_016883010.1
XM_017027522.1 Intron XP_016883011.1

View Full Product Details