Product Details

SNP ID
rs117518271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5632581 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTCCAGAGTCATGCATTGCATCA[A/G]GTTGTCATGTTTCCTTAGTTTTCTT
Phenotype
MIM: 602895 MIM: 608066
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SAFB PubMed Links

Gene Details

Gene
SAFB
Gene Name
scaffold attachment factor B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201338.1 Intron NP_001188267.1
NM_001201339.1 Intron NP_001188268.1
NM_001201340.1 Intron NP_001188269.1
NM_001320571.1 Intron NP_001307500.1
NM_001320572.1 Intron NP_001307501.1
NM_002967.3 Intron NP_002958.2
XM_017027114.1 Intron XP_016882603.1
XM_017027115.1 Intron XP_016882604.1
XM_017027116.1 Intron XP_016882605.1
XM_017027117.1 Intron XP_016882606.1
XM_017027118.1 Intron XP_016882607.1
Gene
SAFB2
Gene Name
scaffold attachment factor B2
There are no transcripts associated with this gene.

View Full Product Details