Product Details

SNP ID
rs112259635
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7005368 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAATTGTGTTTATGAGGCATTCT[C/T]GTTCTATGGAAACAAGGAATGGGAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FLJ25758 PubMed Links

Gene Details

Gene
FLJ25758
Gene Name
microtubule affinity regulating kinase 1 pseudogene
There are no transcripts associated with this gene.

View Full Product Details