Product Details

SNP ID
rs113892276
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10288137 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCTATGCCGGCTGAGCGAGAAA[A/T]AGAGGAATATGAAACAATCTGGGGA
Phenotype
MIM: 147840 MIM: 614088 MIM: 601852
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
ICAM1 PubMed Links

Gene Details

Gene
ICAM1
Gene Name
intercellular adhesion molecule 1
There are no transcripts associated with this gene.

Gene
ICAM4
Gene Name
intercellular adhesion molecule 4 (Landsteiner-Wiener blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039132.2 1042 Nonsense Mutation AAG,TAG K258* NP_001034221.1
NM_001544.4 1042 UTR 3 NP_001535.1
NM_022377.3 1042 UTR 3 NP_071772.1
Gene
ICAM5
Gene Name
intercellular adhesion molecule 5
There are no transcripts associated with this gene.

View Full Product Details