Product Details

SNP ID
rs114810281
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10305136 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGGACACTCCACATCTGCCACA[C/T]AGCCTCTTGGGCTGGACATTTTTCC
Phenotype
MIM: 614585 MIM: 601852 MIM: 611639
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FDX1L PubMed Links

Gene Details

Gene
FDX1L
Gene Name
ferredoxin 1 like
There are no transcripts associated with this gene.

Gene
ICAM5
Gene Name
intercellular adhesion molecule 5
There are no transcripts associated with this gene.

Gene
ZGLP1
Gene Name
zinc finger, GATA-like protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001103167.1 1647 Silent Mutation CTA,CTG L257L NP_001096637.1

View Full Product Details