Product Details

SNP ID
rs117814624
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10334285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCACCGGCACCTCCCGGCTGGAG[C/G]CTTCCTTCAAACACCGCAGCTCGGG
Phenotype
MIM: 146631 MIM: 609950
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ICAM3 PubMed Links

Gene Details

Gene
ICAM3
Gene Name
intercellular adhesion molecule 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320605.1 1853 Missense Mutation GCC,GGC A343G NP_001307534.1
NM_001320606.1 1853 Missense Mutation GCC,GGC A362G NP_001307535.1
NM_001320608.1 1853 Missense Mutation GCC,GGC A178G NP_001307537.1
NM_002162.4 1853 Missense Mutation GCC,GGC A439G NP_002153.2
Gene
RAVER1
Gene Name
ribonucleoprotein, PTB binding 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133452.2 1853 Intron NP_597709.2
XM_011527671.1 1853 Intron XP_011525973.1

View Full Product Details