Product Details

SNP ID
rs112190412
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10420949 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCGCCGAGCGGGAGCGGCAGCCGC[A/G]CCGGCCCATAGAGCGCGCCGATGCC
Phenotype
MIM: 600126
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDE4A PubMed Links

Gene Details

Gene
PDE4A
Gene Name
phosphodiesterase 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111307.1 206 Missense Mutation CAC,CGC H62R NP_001104777.1
NM_001111308.1 206 Intron NP_001104778.1
NM_001111309.1 206 Intron NP_001104779.1
NM_001243121.1 206 Intron NP_001230050.1
NM_006202.2 206 Intron NP_006193.1
XM_005259933.2 206 Intron XP_005259990.1
XM_011528054.1 206 Intron XP_011526356.1
XM_011528055.2 206 Missense Mutation CAC,CGC H62R XP_011526357.1
XM_017026865.1 206 Missense Mutation CAC,CGC H62R XP_016882354.1

View Full Product Details