Product Details

SNP ID
rs114213467
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10702112 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACAGCTTTGCGGTGGGGTTTCCC[G/T]TAGGACAGCGGCGGTTTCCAGATGG
Phenotype
MIM: 603182 MIM: 609615
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
ILF3 PubMed Links

Gene Details

Gene
ILF3
Gene Name
interleukin enhancer binding factor 3
There are no transcripts associated with this gene.

Gene
QTRT1
Gene Name
queuine tRNA-ribosyltransferase catalytic subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031209.2 Intron NP_112486.1

View Full Product Details