Product Details

SNP ID
rs112797378
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11423996 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATGCAGGCTGTCCTGGATGGTGT[C/G]GTCGGACTGTAGCAGCAGGTGCTCG
Phenotype
MIM: 615956 MIM: 616743
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC151 PubMed Links

Gene Details

Gene
CCDC151
Gene Name
coiled-coil domain containing 151
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302453.1 1079 Missense Mutation CAC,GAC H279D NP_001289382.1
NM_001302454.1 1079 Missense Mutation CAC,GAC H273D NP_001289383.1
NM_145045.4 1079 Missense Mutation CAC,GAC H333D NP_659482.3
XM_017026241.1 1079 Missense Mutation CCA,CGA P313R XP_016881730.1
Gene
RGL3
Gene Name
ral guanine nucleotide dissociation stimulator like 3
There are no transcripts associated with this gene.

View Full Product Details