Product Details

SNP ID
rs111926934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12973314 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGCCTGTCAAGGTGGGGTGGAA[A/G]TGGACAGGTGATTATCCTCGCCACT
Phenotype
MIM: 609068
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DAND5 PubMed Links

Gene Details

Gene
DAND5
Gene Name
DAN domain BMP antagonist family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152654.2 Intron NP_689867.1

View Full Product Details