Product Details

SNP ID
rs117581914
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9126048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGTATCCTAGATATTAGTTTCCT[C/G]AAAGCCTTCAACATGTCCTTGTTTC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR7G3 PubMed Links

Gene Details

Gene
OR7G3
Gene Name
olfactory receptor family 7 subfamily G member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001958.1 903 Missense Mutation TTC,TTG F301L NP_001001958.1

View Full Product Details