Product Details

SNP ID
rs116775054
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:13956192 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGACGAGAACCACCGTGACATCTA[C/T]GTCAGCACCGTGGCCGTGCCACCGC
Phenotype
MIM: 600006
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCAF15 PubMed Links

Gene Details

Gene
DCAF15
Gene Name
DDB1 and CUL4 associated factor 15
There are no transcripts associated with this gene.

Gene
PODNL1
Gene Name
podocan like 1
There are no transcripts associated with this gene.

Gene
RFX1
Gene Name
regulatory factor X1
There are no transcripts associated with this gene.

View Full Product Details