Product Details

SNP ID
rs113811779
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16680078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTACATCCTGGCTGATCTGCTCCT[C/T]GGGGAGCCACTGATCGATTACTTCA
Phenotype
MIM: 611235
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM38A PubMed Links

Gene Details

Gene
TMEM38A
Gene Name
transmembrane protein 38A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024074.2 310 Silent Mutation CTC,CTT L73L NP_076979.1

View Full Product Details