Product Details

SNP ID
rs115760264
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16732804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGCAGCGCTATCCCTGGCTAACC[A/G]GGGATGTTTCTGCCTGCACATTTCC
Phenotype
MIM: 616250
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NWD1 PubMed Links

Gene Details

Gene
NWD1
Gene Name
NACHT and WD repeat domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007525.3 Intron NP_001007526.3
NM_001290355.1 Intron NP_001277284.1
XM_011527929.1 Intron XP_011526231.1
XM_011527930.2 Intron XP_011526232.1
XM_011527931.2 Intron XP_011526233.1
XM_011527934.1 Intron XP_011526236.1
XM_011527935.1 Intron XP_011526237.1
XM_017026669.1 Intron XP_016882158.1

View Full Product Details