Product Details

SNP ID
rs115219027
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17601615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTTGTGTGGAGGGGAAGCCTCCC[C/T]GGACTGCTTGGCCGGTTTGTGGGGG
Phenotype
MIM: 609894
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UNC13A PubMed Links

Gene Details

Gene
UNC13A
Gene Name
unc-13 homolog A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080421.2 9693 UTR 3 NP_001073890.2
XM_011527810.2 9693 UTR 3 XP_011526112.1
XM_011527811.2 9693 UTR 3 XP_011526113.1
XM_017026502.1 9693 UTR 3 XP_016881991.1

View Full Product Details