Product Details

SNP ID
rs116471481
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9527670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATATTAAGGCTTTTGGACAATCTA[C/T]AAACGTTTTTGCTTATTGCTTACTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF426 PubMed Links

Gene Details

Gene
ZNF426
Gene Name
zinc finger protein 426
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300883.2 2593 UTR 3 NP_001287812.1
NM_001318055.1 2593 Intron NP_001304984.1
NM_001318056.1 2593 Intron NP_001304985.1
NM_024106.2 2593 Intron NP_077011.1
XM_017027293.1 2593 UTR 3 XP_016882782.1
XM_017027294.1 2593 UTR 3 XP_016882783.1

View Full Product Details